Learn More
Medchemexpress LLC Delafloxacin-d5 | 189279-58-1 | 99.8% | 445.79 | 1 MG
SDP
Catalog No. 5000362532
Encompass_Preferred
Delafloxacin-d5 is a deuterium labeled version of Delafloxacin, a broad-spectrum fluoroquinolone antibiotic. It exhibits activity against drug-resistant bacteria such as Staphylococcus aureus, Streptococcus pneumoniae, and Klebsiella pneumonia.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-62532 Supplier Medchemexpress LLC Supplier No. HY14814S1MG
Delafloxacin-d5 is a deuterium labeled version of Delafloxacin, a broad-spectrum fluoroquinolone antibiotic. It exhibits activity against drug-resistant bacteria such as Staphylococcus aureus, Streptococcus pneumoniae, and Klebsiella pneumonia.
  • Can be used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS