Learn More
Medchemexpress LLC HY-14814A 10mg Medchemexpress, Delafloxacin (meglumine) CAS:352458-37-8 Purity:>98%
SDP
Catalog No. 501932708
Encompass_Preferred
Medchemexpress, HY-14814A 10mg Delafloxacin (meglumine) CAS:352458-37-8 Delafloxacin meglumine (ABT492 meglumine; RX-3341 meglumine; WQ-3034 meglumine) is a broad-spectrum fluoroquinolone antibiotic. Delafloxacin has a broad spectrum of activity that includes drug-resistant Staphylococcus aureus, Streptococcus pneumoniae, and Klebsiella pneumonia [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2708 Supplier Medchemexpress LLC Supplier No. HY14814A10MG
Medchemexpress, HY-14814A 10mg Delafloxacin (meglumine) CAS:352458-37-8 Delafloxacin meglumine (ABT492 meglumine; RX-3341 meglumine; WQ-3034 meglumine) is a broad-spectrum fluoroquinolone antibiotic. Delafloxacin has a broad spectrum of activity that includes drug-resistant Staphylococcus aureus, Streptococcus pneumoniae, and Klebsiella pneumonia [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.