Learn More
Selleck Chemical LLC Delafloxacin Meglumine S4488-5mg
SDP
Catalog No. 503110244
Encompass_Preferred
Delafloxacin (ABT-492 RX-3341 WQ-3034 Baxdela) meglumine is a broad-spectrum fluoroquinolone antibiotic against multiple pathogens
Catalog No. 50-311-0244 Supplier Selleck Chemical LLC Supplier No. S44885MG
Delafloxacin (ABT-492 RX-3341 WQ-3034 Baxdela) meglumine is a broad-spectrum fluoroquinolone antibiotic against multiple pathogens