Learn More
Cayman Chemical DelafloxacInmeglumIn 100mg
SDP
Catalog No. 502938937
Encompass_Preferred
A fluoroquinolone antibiotic; active against various bacteria in vitro (MICs = S. aureus and S. pneumoniae lung infection from 3-15.5 mg/kg
Catalog No. 50-293-8937 Supplier Cayman Chemical Supplier No. 27935100 MG
A fluoroquinolone antibiotic; active against various bacteria in vitro (MICs = S. aureus and S. pneumoniae lung infection from 3-15.5 mg/kg