Learn More
Medchemexpress LLC HY-10846 10mg Medchemexpress, Delamanid CAS:681492-22-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1084610MG
Encompass_Preferred
Medchemexpress, HY-10846 10mg Delamanid CAS:681492-22-8 Delamanid, a newer mycobacterial cell wall synthesis inhibitor, inhibits the synthesisi of mucolic acids. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8785