Learn More
Selleck Chemical LLC Delamanid-5mg
SDP
Catalog No. 502389680
Encompass_Preferred
Delamanid (OPC-67683) is a new anti-tuberculosis drug with an excellent intracellular bactericidal activity and a high accumulation rate.
Catalog No. 50-238-9680 Supplier Selleck Chemical LLC Supplier No. S50075MG
Delamanid (OPC-67683) is a new anti-tuberculosis drug with an excellent intracellular bactericidal activity and a high accumulation rate.