Learn More
Selleck Chemical LLC Delanzomib CEP-18770-5mg
SDP
Supplier:  Selleck Chemical LLC S11575MG
Encompass_Preferred
Delanzomib (CEP-18770) is an orally active inhibitor of the chymotrypsin-like activity of proteasome with IC50 of 3.8 nM, with only marginal inhibition of the tryptic and peptidylglutamyl activities of the proteosome. Phase 1/2.
Catalog No. 50-238-9783