Learn More
TARGETMOL CHEMICALS INC DELAPRIL HYDROCHLORIDE 25MG
SDP
Catalog No. 502576965
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Delapril hydrochloride (indalapril) is a prodrug; it is converted into two active metabolites 5-hydroxy delapril diacid and delapril diacid. These metabolites bind completely to and inhibit angiotensin-converting enzyme (ACE) hence blocking angiotensin I to angiotensin II conversion. purity: 99%
Catalog No. 50-257-6965 Supplier TARGETMOL CHEMICALS INC Supplier No. T328125MG
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Delapril hydrochloride (indalapril) is a prodrug; it is converted into two active metabolites 5-hydroxy delapril diacid and delapril diacid. These metabolites bind completely to and inhibit angiotensin-converting enzyme (ACE) hence blocking angiotensin I to angiotensin II conversion. purity: 99%