Learn More
Selleck Chemical LLC Delapril Hydrochloride
SDP
Catalog No. 5000003085
Encompass_Preferred
Delapril Hydrochloride (Alindapril) is the hydrochloride salt form of delapril which is a lipophilic non-sulfhydryl angiotensin-converting enzyme (ACE) inhibitor with antihypertensive activity
Catalog No. 50-000-03085 Supplier Selleck Chemical LLC Supplier No. S57285MG
Delapril Hydrochloride (Alindapril) is the hydrochloride salt form of delapril which is a lipophilic non-sulfhydryl angiotensin-converting enzyme (ACE) inhibitor with antihypertensive activity