Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Delapril hydrochloride | 83435-67-0 | 98.0% | 489.00 g·mol⁻¹ | C26H33ClN2O5 | 1 ML
SDP

Catalog No. 5000241133
Encompass_Preferred

Delapril hydrochloride is the hydrochloride salt of delapril, an angiotensin-converting enzyme (ACE) inhibitor used in cardiovascular research and preclinical studies. The compound is provided both as a ready-to-use 10 mM solution in DMSO (1 mL) and in solid quantities. Typical reported purity is 98.0% and the molecular weight is 489.00 g·mol⁻¹. Store and handle according to supplier safety data.

  • ACE inhibitor used in cardiovascular research.
  • Available as 10 mM solution in DMSO (1 mL) and as solid quantities.
  • High reported purity, 98.0%.
  • Molecular weight 489.00 g·mol⁻¹.
  • CAS number 83435-67-0 for unambiguous identification.
  • Store solid at 4°C; store solution frozen for long-term stability.

Catalog No. 50-002-41133 Supplier Medchemexpress LLC Supplier No. HY10733710MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Delapril hydrochloride is the hydrochloride salt of delapril, an angiotensin-converting enzyme (ACE) inhibitor used in cardiovascular research and preclinical studies. The compound is provided both as a ready-to-use 10 mM solution in DMSO (1 mL) and in solid quantities. Typical reported purity is 98.0% and the molecular weight is 489.00 g·mol⁻¹. Store and handle according to supplier safety data.

  • ACE inhibitor used in cardiovascular research.
  • Available as 10 mM solution in DMSO (1 mL) and as solid quantities.
  • High reported purity, 98.0%.
  • Molecular weight 489.00 g·mol⁻¹.
  • CAS number 83435-67-0 for unambiguous identification.
  • Store solid at 4°C; store solution frozen for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.