Learn More
Medchemexpress LLC Delapril hydrochloride | 83435-67-0 | MFCD00884619 | 98.0% | 489.00 | C26H33ClN2O5 | 50 MG
SDP
Catalog No. 5000240958
Encompass_Preferred
Delapril hydrochloride is the hydrochloride salt of delapril, a lipophilic angiotensin-converting enzyme (ACE) inhibitor commonly used in cardiovascular research. Supplied for research use only, the compound is characterized for identity and analytical purity to support experimental studies.
  • Hydrochloride salt form for improved solubility and stability.
  • High purity: 98.0% (NMR).
  • Molecular formula C26H33ClN2O5; molecular weight 489.00.
  • Available in solid quantities including 50 mg.
  • Analytical documentation available: COA and SDS.
  • Intended for in vitro and preclinical ACE inhibition studies.
Catalog No. 50-002-40958 Supplier Medchemexpress LLC Supplier No. HY10733750MG
Delapril hydrochloride is the hydrochloride salt of delapril, a lipophilic angiotensin-converting enzyme (ACE) inhibitor commonly used in cardiovascular research. Supplied for research use only, the compound is characterized for identity and analytical purity to support experimental studies.
  • Hydrochloride salt form for improved solubility and stability.
  • High purity: 98.0% (NMR).
  • Molecular formula C26H33ClN2O5; molecular weight 489.00.
  • Available in solid quantities including 50 mg.
  • Analytical documentation available: COA and SDS.
  • Intended for in vitro and preclinical ACE inhibition studies.