Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Delapril (hydrochloride) | 83435-67-0 | MFCD00884619 | 98.0% | 489.00 g·mol⁻¹ | C26H33ClN2O5 | 100 MG
SDP

Catalog No. 5000241499
Encompass_Preferred

Delapril hydrochloride is the hydrochloride salt of delapril, a lipophilic angiotensin-converting enzyme (ACE) inhibitor used in preclinical research on hypertension and cardiovascular pharmacology. The compound is supplied as a solid research reagent with confirmed identity and analytical data.

  • Research reagent for ACE inhibition and cardiovascular studies.
  • CAS number 83435-67-0.
  • Molecular formula C26H33ClN2O5; molecular weight 489.00 g·mol⁻¹.
  • Purity 98.0% (NMR) as stated on COA.
  • Physical appearance solid.
  • Recommended storage 4°C; in solvent -80°C for 6 months, -20°C for 1 month (sealed).
  • Packaged as 100 mg research quantity.

Catalog No. 50-002-41499 Supplier Medchemexpress LLC Supplier No. HY107337100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Delapril hydrochloride is the hydrochloride salt of delapril, a lipophilic angiotensin-converting enzyme (ACE) inhibitor used in preclinical research on hypertension and cardiovascular pharmacology. The compound is supplied as a solid research reagent with confirmed identity and analytical data.

  • Research reagent for ACE inhibition and cardiovascular studies.
  • CAS number 83435-67-0.
  • Molecular formula C26H33ClN2O5; molecular weight 489.00 g·mol⁻¹.
  • Purity 98.0% (NMR) as stated on COA.
  • Physical appearance solid.
  • Recommended storage 4°C; in solvent -80°C for 6 months, -20°C for 1 month (sealed).
  • Packaged as 100 mg research quantity.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.