Learn More
Apexbio Technology LLC Delavirdine mesylate 147221-93-0 200mg
SDP
Catalog No. 5000089226
Encompass_Preferred
Delavirdine mesylate (CAS 147221-93-0) is a small-molecule inhibitor targeting HIV-1 reverse transcriptase It is designed to selectively inhibit the enzymatic activity of HIV-1 reverse transcriptase thereby disrupting viral DNA synthesis and impeding HIV replication Delavirdine mesylate exerts its biological activity primarily through direct binding to an allosteric site on HIV-1 reverse transcriptase In biochemical studies delavirdine mesylate demonstrates inhibitory activity against HIV-1 reverse transcriptase with an IC50 value of 0 26 M Based on these pharmacological properties delavirdine mesylate holds research potential in antiviral assays HIV resistance studies and mechanistic investigations into NNRTI-related enzyme inhibition
Catalog No. 50-000-89226 Supplier Apexbio Technology LLC Supplier No. B1050200
Delavirdine mesylate (CAS 147221-93-0) is a small-molecule inhibitor targeting HIV-1 reverse transcriptase It is designed to selectively inhibit the enzymatic activity of HIV-1 reverse transcriptase thereby disrupting viral DNA synthesis and impeding HIV replication Delavirdine mesylate exerts its biological activity primarily through direct binding to an allosteric site on HIV-1 reverse transcriptase In biochemical studies delavirdine mesylate demonstrates inhibitory activity against HIV-1 reverse transcriptase with an IC50 value of 0 26 M Based on these pharmacological properties delavirdine mesylate holds research potential in antiviral assays HIV resistance studies and mechanistic investigations into NNRTI-related enzyme inhibition