Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Delavirdine mesylate | 147221-93-0 | 98.0% | 552.67 g/mol | C23H32N6O6S2 | 5 MG
SDP

Catalog No. 5000242057
Encompass_Preferred

Delavirdine mesylate is the mesylate salt of delavirdine, a non-nucleoside reverse transcriptase inhibitor (NNRTI) used in research to study HIV-1 reverse transcriptase inhibition. Supplied as a solid with reported molecular formula C23H32N6O6S2 and molecular weight 552.67 g/mol, this research chemical is provided with batch analytical data and safety documentation for laboratory use only.

  • Non-nucleoside reverse transcriptase inhibitor for HIV-1 research.
  • Reported purity 98.0% (LCMS) for the supplied batch.
  • Molecular formula C23H32N6O6S2; molecular weight 552.67 g/mol.
  • Available in small pack sizes suitable for assay development and analytical studies.
  • Accompanied by batch COA and SDS for quality and safety information.

Catalog No. 50-002-42057 Supplier Medchemexpress LLC Supplier No. HY10571A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Delavirdine mesylate is the mesylate salt of delavirdine, a non-nucleoside reverse transcriptase inhibitor (NNRTI) used in research to study HIV-1 reverse transcriptase inhibition. Supplied as a solid with reported molecular formula C23H32N6O6S2 and molecular weight 552.67 g/mol, this research chemical is provided with batch analytical data and safety documentation for laboratory use only.

  • Non-nucleoside reverse transcriptase inhibitor for HIV-1 research.
  • Reported purity 98.0% (LCMS) for the supplied batch.
  • Molecular formula C23H32N6O6S2; molecular weight 552.67 g/mol.
  • Available in small pack sizes suitable for assay development and analytical studies.
  • Accompanied by batch COA and SDS for quality and safety information.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.