Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Delavirdine mesylate | 147221-93-0 | MFCD00866948 | 552.67 | C23H32N6O6S2 | 50 MG
SDP

Catalog No. 5000241551
Encompass_Preferred

Delavirdine mesylate is the mesylate salt of delavirdine, a non-nucleoside reverse transcriptase inhibitor (NNRTI) provided for research and analytical applications. It is supplied as a solid with defined composition and molecular weight for use in in vitro studies of HIV-1 reverse transcriptase. Not for human or clinical use.

  • CAS registry number: 147221-93-0.
  • Molecular formula: C23H32N6O6S2.
  • Molecular weight: 552.67.
  • Physical form: solid; appearance white to yellow.
  • Available pack sizes: 5 mg, 10 mg, 50 mg, 100 mg, 200 mg.
  • Intended use: for research and analytical applications only; not for human use.

Catalog No. 50-002-41551 Supplier Medchemexpress LLC Supplier No. HY10571A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Delavirdine mesylate is the mesylate salt of delavirdine, a non-nucleoside reverse transcriptase inhibitor (NNRTI) provided for research and analytical applications. It is supplied as a solid with defined composition and molecular weight for use in in vitro studies of HIV-1 reverse transcriptase. Not for human or clinical use.

  • CAS registry number: 147221-93-0.
  • Molecular formula: C23H32N6O6S2.
  • Molecular weight: 552.67.
  • Physical form: solid; appearance white to yellow.
  • Available pack sizes: 5 mg, 10 mg, 50 mg, 100 mg, 200 mg.
  • Intended use: for research and analytical applications only; not for human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.