Learn More
Cayman Chemical Delavirdine (mesylate), Purity: ≥98%, CAS: 147221-93-0, 10 mg
SDP
Catalog No. 502934138
Encompass_Preferred
A NNRTI; selectively inhibits HIV-1 reverse transcriptase over DNA polymerase α and δ in vitro (IC50s = 0.26, 440, and >550 μM, respectively); blocks replication of 25 primary HIV-1 isolates (mean ED50 = 0.066 μM); inhibits growth of L. infantum promastigotes (IC50 = 26.1 μM)
Catalog No. 50-293-4138 Supplier Cayman Chemical Supplier No. 2402610 MG
A NNRTI; selectively inhibits HIV-1 reverse transcriptase over DNA polymerase α and δ in vitro (IC50s = 0.26, 440, and >550 μM, respectively); blocks replication of 25 primary HIV-1 isolates (mean ED50 = 0.066 μM); inhibits growth of L. infantum promastigotes (IC50 = 26.1 μM)