Learn More
Waters Corp Delay Coil, 2–8 mL, 100:1 Ratio, 0.020 × 250
Catalog No. 50792891 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Waters Delay Coil is a spare part used in LC systems to control mobile phase delay time, ensuring precise analytical conditions. It offers a capacity of 2–8 mL and a 100:1 ratio, and with its 0.020 × 250 specification, it is suitable for high-performance liquid chromatography (HPLC) applications.
  • Coil for adjusting delay time in LC systems
  • Stable flow control and improved reproducibility
  • Easy replacement due to its spare part kit form
  • Optimized compatibility with LC systems
Catalog No. 50-792-891 Supplier Waters Corp Supplier No. 700003604
The Waters Delay Coil is a spare part used in LC systems to control mobile phase delay time, ensuring precise analytical conditions. It offers a capacity of 2–8 mL and a 100:1 ratio, and with its 0.020 × 250 specification, it is suitable for high-performance liquid chromatography (HPLC) applications.
  • Coil for adjusting delay time in LC systems
  • Stable flow control and improved reproducibility
  • Easy replacement due to its spare part kit form
  • Optimized compatibility with LC systems