Learn More
TARGETMOL CHEMICALS INC DELEQUAMINE 5MG
SDP
Catalog No. 502576448
Encompass_Preferred
Also available in 1mg 10mg 25mg 50mg 100mg 500mg and bulk. Please contact Fisher for quotes. Delequamine (RS 15385-197) is a selective and potent TNIK inhibitor (IC50: 8 nM).TINK-IN-1 inhibits colorectal cancer cell viability and can be used to study mental retardation. purity: 98%
Catalog No. 50-257-6448 Supplier TARGETMOL CHEMICALS INC Supplier No. T680515MG
Also available in 1mg 10mg 25mg 50mg 100mg 500mg and bulk. Please contact Fisher for quotes. Delequamine (RS 15385-197) is a selective and potent TNIK inhibitor (IC50: 8 nM).TINK-IN-1 inhibits colorectal cancer cell viability and can be used to study mental retardation. purity: 98%