Learn More
Cayman Chemical Delgocitinib, Purity: ≥98%, CAS: 1263774-59-9, 10 mg
SDP
Catalog No. 502915902
Encompass_Preferred
An inhibitor of JAK1, JAK2, JAK3, and TYK2 (IC50s = 2.8, 2.6, 13, and 58 nM, respectively); is selective for JAK1, JAK2, JAK3, and TYK2 over LCK (IC50 = 5,800 nM); inhibits IL-31-induced increases in STAT3 phosphorylation levels in A549 cells (IC50 = 25 nM); reduces IL-31-induced scratching in mice in a dose-dependent manner
Catalog No. 50-291-5902 Supplier Cayman Chemical Supplier No. 3830710 MG
An inhibitor of JAK1, JAK2, JAK3, and TYK2 (IC50s = 2.8, 2.6, 13, and 58 nM, respectively); is selective for JAK1, JAK2, JAK3, and TYK2 over LCK (IC50 = 5,800 nM); inhibits IL-31-induced increases in STAT3 phosphorylation levels in A549 cells (IC50 = 25 nM); reduces IL-31-induced scratching in mice in a dose-dependent manner