Learn More
Medchemexpress LLC HY-109053 1mg , Delgocitinib CAS:1263774-59-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1090531MG
Encompass_Preferred
HY-109053 1mg Delgocitinib CAS: 1263774-59-9 Delgocitinib (JTE-052) is a specific JAK inhibitor with IC50s of 2.8, 2.6, 13 and 58 nM for JAK1, JAK2, JAK3 and Tyk2, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-225-9925