Learn More
Medchemexpress LLC HY-109053 5mg Medchemexpress, Delgocitinib CAS:1263774-59-9 Purity:>98%
SDP
Catalog No. 501968660
Encompass_Preferred
Medchemexpress, HY-109053 5mg Delgocitinib CAS:1263774-59-9 Delgocitinib is a novel and specific JAK inhibitor with IC50s of 2.8, 2.6, 13 and 58 nM for JAK1, JAK2, JAK3 and Tyk2, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8660 Supplier Medchemexpress LLC Supplier No. HY1090535MG
Medchemexpress, HY-109053 5mg Delgocitinib CAS:1263774-59-9 Delgocitinib is a novel and specific JAK inhibitor with IC50s of 2.8, 2.6, 13 and 58 nM for JAK1, JAK2, JAK3 and Tyk2, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.