Learn More
Selleck Chemical LLC Delgocitinib S8802-2MG
SDP
Catalog No. 5000023658
Encompass_Preferred
Delgocitinib (JTE-052) is a specific JAK inhibitor with IC50s of 2 8 2 6 13 and 58 nM for JAK1 JAK2 JAK3 and Tyk2 respectively
Catalog No. 50-000-23658 Supplier Selleck Chemical LLC Supplier No. S88022MG
Delgocitinib (JTE-052) is a specific JAK inhibitor with IC50s of 2 8 2 6 13 and 58 nM for JAK1 JAK2 JAK3 and Tyk2 respectively