Learn More
MSC Kimtech Kimwipes Delicate Task Wipes, Pop-Up Box, Clean Room, Dry, 12.4 x 11.23 in
Catalog No. 502837095
Encompass
These delicate task wipes provide clean room quality for sensitive applications. Presented in a convenient pop-up box, each single-ply, unscented, white wipe is ideal for a variety of laboratory and delicate cleaning requirements.
  • Clean room wipe type
  • Flat sheet form
  • Dry wipe style
  • Pop-up box packaging
  • Sheet length: 12.4 in
  • Sheet width: 11.23 in
  • White wipe color
  • Unscented
  • One ply
  • 198 sheets per box
Catalog No. 50-283-7095 Supplier MSC Supplier No. 67690107
These delicate task wipes provide clean room quality for sensitive applications. Presented in a convenient pop-up box, each single-ply, unscented, white wipe is ideal for a variety of laboratory and delicate cleaning requirements.
  • Clean room wipe type
  • Flat sheet form
  • Dry wipe style
  • Pop-up box packaging
  • Sheet length: 12.4 in
  • Sheet width: 11.23 in
  • White wipe color
  • Unscented
  • One ply
  • 198 sheets per box