Learn More
Chemglass Life Sciences Delivery Adapter Only, 24/40, Airfree, Schlenk
SDP
Catalog No. AF072004
Encompass_Preferred
Delivery Adapter Only, 24/40, Airfree, Schlenk. Delivery adapter for use with item AF072002. Delivery adapter is fitted w/a 12/5 spherical joint for flexibility & a 0-4mm Chem-Vac™ Chem-Cap valve for purging of the receiving vessel. Top outer joint is a 24/40 while the uptake tube is a 24/40 inner joint. Delivery adapter has a 14/20 inner joint & is supplied w/the complete apparatus along w/a CG15002 Stainless Steel Pinch Clamp.
Catalog No. AF072004 Supplier Chemglass Life Sciences Supplier No. AF072004
Delivery Adapter Only, 24/40, Airfree, Schlenk. Delivery adapter for use with item AF072002. Delivery adapter is fitted w/a 12/5 spherical joint for flexibility & a 0-4mm Chem-Vac™ Chem-Cap valve for purging of the receiving vessel. Top outer joint is a 24/40 while the uptake tube is a 24/40 inner joint. Delivery adapter has a 14/20 inner joint & is supplied w/the complete apparatus along w/a CG15002 Stainless Steel Pinch Clamp.