Learn More
BURKLE INC Delivery tubes PE for OTALdisposable pump, Ø 0.47 in. (12 mm)
Catalog No. 5000025557
Encompass_Preferred
The HDPE delivery tubes are accessories for the OTAL disposable pumps.
They are made of high quality, high density polyethylene, have excellent chemical resistance and are resistant to a wide range of chemicals.
The tubes can be shortened as required to fit a wide range of requirements.
Catalog No. 50-000-25557 Supplier BURKLE INC Supplier No. 50008000
The HDPE delivery tubes are accessories for the OTAL disposable pumps.
They are made of high quality, high density polyethylene, have excellent chemical resistance and are resistant to a wide range of chemicals.
The tubes can be shortened as required to fit a wide range of requirements.