Learn More
Perkin Elmer US LLC DELL WINDOWS 10 SYSTEM ICP
Catalog No. NC3628272
Encompass_Preferred
  • Dedicated controller for ICP-OES systems
  • Optimizes operational efficiency of instruments
  • Equipped with Windows 10 OS for stable performance
  • Seamless integration with Syngistix for ICP software
  • Facilitates intuitive method development
  • Enables real-time instrument diagnostics
  • Provides comprehensive data review
  • Reduces turnaround times in laboratory environments
  • Enhances analytical confidence for scientists
  • Ensures reliable communication and control over ICP processes
Catalog No. NC3628272 Supplier Perkin Elmer US LLC Supplier No. N0780205
Add to Cart
Add to Cart
Request a Quote
  • Dedicated controller for ICP-OES systems
  • Optimizes operational efficiency of instruments
  • Equipped with Windows 10 OS for stable performance
  • Seamless integration with Syngistix for ICP software
  • Facilitates intuitive method development
  • Enables real-time instrument diagnostics
  • Provides comprehensive data review
  • Reduces turnaround times in laboratory environments
  • Enhances analytical confidence for scientists
  • Ensures reliable communication and control over ICP processes