Learn More
Medchemexpress LLC Delmitide acetate 10mg | 501019-16-5 | 10MG
SDP
Catalog No. 5000244282
Encompass_Preferred
Delmitide (RDP58) acetate is an orally active d-isomer decapeptide with potent anti-inflammatory activity Delmitide acetate inhibits production of TNF- IFN- and interleukin (IL)-12 and up-regulates heme oxygenase 1 activity Delmitide acetate can be used for the research of ulcerative colitis[1 [2
Catalog No. 50-002-44282 Supplier Medchemexpress LLC Supplier No. HY106359A10MG
Delmitide (RDP58) acetate is an orally active d-isomer decapeptide with potent anti-inflammatory activity Delmitide acetate inhibits production of TNF- IFN- and interleukin (IL)-12 and up-regulates heme oxygenase 1 activity Delmitide acetate can be used for the research of ulcerative colitis[1 [2