Learn More
Medchemexpress LLC Delmitide acetate | 501019-16-5 | 99.2% | 1288.62 g/mol | C59H105N17O11.C2H4O2 | 5 MG
SDP
Catalog No. 5000244387
Encompass_Preferred
Delmitide acetate is an orally active D-isomer decapeptide with potent anti-inflammatory activity. It inhibits production of TNF-α, IFN-γ, and IL-12, and up-regulates heme oxygenase-1, making it useful for preclinical studies such as ulcerative colitis and cytokine modulation research.
  • Orally active d-isomer decapeptide.
  • Inhibits pro-inflammatory cytokines, including TNF-α, IFN-γ, and IL-12.
  • Up-regulates heme oxygenase-1 to modulate inflammatory response.
  • Suitable for ulcerative colitis and inflammation studies.
  • Provided as an acetate salt in small research quantities.
  • High purity suitable for preclinical research.
Catalog No. 50-002-44387 Supplier Medchemexpress LLC Supplier No. HY106359A5MG
Delmitide acetate is an orally active D-isomer decapeptide with potent anti-inflammatory activity. It inhibits production of TNF-α, IFN-γ, and IL-12, and up-regulates heme oxygenase-1, making it useful for preclinical studies such as ulcerative colitis and cytokine modulation research.
  • Orally active d-isomer decapeptide.
  • Inhibits pro-inflammatory cytokines, including TNF-α, IFN-γ, and IL-12.
  • Up-regulates heme oxygenase-1 to modulate inflammatory response.
  • Suitable for ulcerative colitis and inflammation studies.
  • Provided as an acetate salt in small research quantities.
  • High purity suitable for preclinical research.