Learn More
Medchemexpress LLC Delmitide acetate | 501019-16-5 | 99.2% | 1288.62 g/mol | C61H109N17O13 | 25 MG
SDP
Catalog No. 5000242257
Encompass_Preferred
Delmitide acetate is an orally active D-isomer decapeptide research compound with reported anti-inflammatory activity. It inhibits production of TNF-α, IFN-γ, and IL-12, and up-regulates heme oxygenase 1. The compound is used in preclinical studies of inflammatory disorders such as ulcerative colitis.
Catalog No. 50-002-42257 Supplier Medchemexpress LLC Supplier No. HY106359A25MG
Delmitide acetate is an orally active D-isomer decapeptide research compound with reported anti-inflammatory activity. It inhibits production of TNF-α, IFN-γ, and IL-12, and up-regulates heme oxygenase 1. The compound is used in preclinical studies of inflammatory disorders such as ulcerative colitis.