Learn More
Medchemexpress LLC Delmitide acetate | 501019-16-5 | 99.2% | 1288.62 g/mol | C61H109N17O13 | 50 MG
SDP
Catalog No. 5000244362
Encompass_Preferred
Delmitide acetate is an orally active D-isomer decapeptide with potent anti-inflammatory activity. It inhibits production of TNF-α, IFN-γ, and interleukin-12, and up-regulates heme oxygenase-1 activity. It is provided for research use in studies of inflammatory diseases such as ulcerative colitis.
  • Orally active d-isomer decapeptide.
  • Potent inhibition of proinflammatory cytokines (TNF-α, IFN-γ, IL-12).
  • Up-regulates heme oxygenase-1 activity.
  • Suitable for inflammatory disease research applications.
  • Supplied as an acetate salt with reported high purity.
Catalog No. 50-002-44362 Supplier Medchemexpress LLC Supplier No. HY106359A50MG
Delmitide acetate is an orally active D-isomer decapeptide with potent anti-inflammatory activity. It inhibits production of TNF-α, IFN-γ, and interleukin-12, and up-regulates heme oxygenase-1 activity. It is provided for research use in studies of inflammatory diseases such as ulcerative colitis.
  • Orally active d-isomer decapeptide.
  • Potent inhibition of proinflammatory cytokines (TNF-α, IFN-γ, IL-12).
  • Up-regulates heme oxygenase-1 activity.
  • Suitable for inflammatory disease research applications.
  • Supplied as an acetate salt with reported high purity.