Learn More
Bellco Glass, Inc. Delong Fernbach Flask, 2800mL No Baffles
SDP
Catalog No. 501945550
Encompass_Preferred
Manufactured from Type 1 Class A Borosilicate Glass. Ideal for culturing organisms requiring a large surface to volume. DeLong style neck and uses 38mm stainless steel closure (sold separately). MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSizes: 2800mlOD x OAH: 205 x 250mmCase Quantity: 3
Catalog No. 50-194-5550 Supplier Bellco Glass, Inc. Supplier No. 255002800
Manufactured from Type 1 Class A Borosilicate Glass. Ideal for culturing organisms requiring a large surface to volume. DeLong style neck and uses 38mm stainless steel closure (sold separately). MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSizes: 2800mlOD x OAH: 205 x 250mmCase Quantity: 3