Learn More
Bellco Glass, Inc. DeLong Flask Complete, 125mL
SDP
Catalog No. 501945453
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Ideal for growing, storing and mixing of aerobic and anaerobic cultures. Graduated to approximate volumes. MADE IN THE USA Size: 125ml Closure: 25 Dimensions: 66x136mm
Catalog No. 50-194-5453 Supplier Bellco Glass, Inc. Supplier No. 251000125
Manufactured from type 1 class a Borosilicate glass. Ideal for growing, storing and mixing of aerobic and anaerobic cultures. Graduated to approximate volumes. MADE IN THE USA Size: 125ml Closure: 25 Dimensions: 66x136mm