Learn More
Bellco Glass, Inc. DeLong Flask Complete, 250mL
SDP
Supplier:  Bellco Glass, Inc. 251000250
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Ideal for growing, storing and mixing of aerobic and anaerobic cultures. Graduated to approximate volumes. MADE IN THE USA Size: 250ml Closure: 38mm Dimensions: 80x160mm
Catalog No. 50-194-5454