Learn More
Bellco Glass, Inc. DeLong Flask Complete, 25mL
SDP
Catalog No. 501945451
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Ideal for growing, storing and mixing of aerobic and anaerobic cultures. Graduated to approximate volumes. MADE IN THE USA Size: 25ml Closure: 18mm Dimensions: 40x96mm
Catalog No. 50-194-5451 Supplier Bellco Glass, Inc. Supplier No. 251000025
Manufactured from type 1 class a Borosilicate glass. Ideal for growing, storing and mixing of aerobic and anaerobic cultures. Graduated to approximate volumes. MADE IN THE USA Size: 25ml Closure: 18mm Dimensions: 40x96mm