Learn More
Bellco Glass, Inc. DeLong Flask Complete, 300ml
SDP
Catalog No. 501945455
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Ideal for growing, storing and mixing of aerobic and anaerobic cultures. Graduated to approximate volumes. MADE IN THE USA Size: 300ml Closure: 38 Dimensions: 88x170mm
Catalog No. 50-194-5455 Supplier Bellco Glass, Inc. Supplier No. 251000300
Manufactured from type 1 class a Borosilicate glass. Ideal for growing, storing and mixing of aerobic and anaerobic cultures. Graduated to approximate volumes. MADE IN THE USA Size: 300ml Closure: 38 Dimensions: 88x170mm