Learn More
Bellco Glass, Inc. DeLong Flask Complete, 500mL
SDP
Catalog No. 501945456
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Ideal for growing, storing and mixing of aerobic and anaerobic cultures. Graduated to approximate volumes. MADE IN THE USA Size: 500ml Closure: 38 Dimensions: 100x205mm
Catalog No. 50-194-5456 Supplier Bellco Glass, Inc. Supplier No. 251000500
Manufactured from type 1 class a Borosilicate glass. Ideal for growing, storing and mixing of aerobic and anaerobic cultures. Graduated to approximate volumes. MADE IN THE USA Size: 500ml Closure: 38 Dimensions: 100x205mm