Learn More
Bellco Glass, Inc. DeLong Flask Complete, 50mL
SDP
Catalog No. 501945452
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Ideal for growing, storing and mixing of aerobic and anaerobic cultures. Graduated to approximate volumes. MADE IN THE USA Size: 50ml Closure: 18 Dimensions: 50x105mm
Catalog No. 50-194-5452 Supplier Bellco Glass, Inc. Supplier No. 251000050
Manufactured from type 1 class a Borosilicate glass. Ideal for growing, storing and mixing of aerobic and anaerobic cultures. Graduated to approximate volumes. MADE IN THE USA Size: 50ml Closure: 18 Dimensions: 50x105mm