Learn More
Bellco Glass, Inc. DeLong Flask Complete, 6L
SDP
Catalog No. 501945461
Encompass_Preferred
Manufactured from type 1 class a Borosilicate glass. Ideal for growing, storing and mixing of aerobic and anaerobic cultures. Graduated to approximate volumes. MADE IN THE USA Size: 6l Closure: 38 Dimensions: 235x410mm
Catalog No. 50-194-5461 Supplier Bellco Glass, Inc. Supplier No. 251006000
Manufactured from type 1 class a Borosilicate glass. Ideal for growing, storing and mixing of aerobic and anaerobic cultures. Graduated to approximate volumes. MADE IN THE USA Size: 6l Closure: 38 Dimensions: 235x410mm