Learn More
Bellco Glass, Inc. DeLong Flask Only,125mL
SDP
Catalog No. 501945464
Encompass_Preferred
Manufactured for type 1 class a Borosilicate glass. Ideal for growing, storing, and mixing of aerobic and anaerobic cultures. Graduated to approximate volumes.. MADE IN THE USA Size: 125ml Closure: 25 Dimensions: 66x136mm Stainless steel closure only sold separately
Catalog No. 50-194-5464 Supplier Bellco Glass, Inc. Supplier No. 251100125
Manufactured for type 1 class a Borosilicate glass. Ideal for growing, storing, and mixing of aerobic and anaerobic cultures. Graduated to approximate volumes.. MADE IN THE USA Size: 125ml Closure: 25 Dimensions: 66x136mm Stainless steel closure only sold separately