Learn More
Bellco Glass, Inc. DeLong Flask Only,250mL
SDP
Catalog No. 501945465
Encompass_Preferred
Manufactured for type 1 class a Borosilicate glass. Ideal for growing, storing, and mixing of aerobic and anaerobic cultures. Graduated to approximate volumes.. MADE IN THE USA Size: 250ml Closure: 38 Dimensions: 80x160 Stainless steel closure or Kaput closure sold separately
Catalog No. 50-194-5465 Supplier Bellco Glass, Inc. Supplier No. 251100250
Manufactured for type 1 class a Borosilicate glass. Ideal for growing, storing, and mixing of aerobic and anaerobic cultures. Graduated to approximate volumes.. MADE IN THE USA Size: 250ml Closure: 38 Dimensions: 80x160 Stainless steel closure or Kaput closure sold separately