Learn More
Bellco Glass, Inc. DeLong Flask Only,25mL
SDP
Catalog No. 501945462
Encompass_Preferred
Manufactured for type 1 class a Borosilicate glass. Ideal for growing, storing, and mixing of aerobic and anaerobic cultures. Graduated to approximate volumes.. MADE IN THE USA Size: 25ml Closure: 18 Dimensions: 40x96mm Stainless steel closure or Kaput closure sold separately
Catalog No. 50-194-5462 Supplier Bellco Glass, Inc. Supplier No. 251100025
Manufactured for type 1 class a Borosilicate glass. Ideal for growing, storing, and mixing of aerobic and anaerobic cultures. Graduated to approximate volumes.. MADE IN THE USA Size: 25ml Closure: 18 Dimensions: 40x96mm Stainless steel closure or Kaput closure sold separately