Learn More
Bellco Glass, Inc. DeLong Flask Only,2L
SDP
Catalog No. 501945469
Encompass_Preferred
Manufactured for type 1 class a Borosilicate glass. Ideal for growing, storing, and mixing of aerobic and anaerobic cultures. Graduated to approximate volumes.. MADE IN THE USA Size: 2l Closure: 38 Dimensions: 158x295 Stainless steel closure or Kaput closure sold separately
Catalog No. 50-194-5469 Supplier Bellco Glass, Inc. Supplier No. 251102000
Manufactured for type 1 class a Borosilicate glass. Ideal for growing, storing, and mixing of aerobic and anaerobic cultures. Graduated to approximate volumes.. MADE IN THE USA Size: 2l Closure: 38 Dimensions: 158x295 Stainless steel closure or Kaput closure sold separately