Learn More
Bellco Glass, Inc. DeLong Flask Only,50mL
SDP
Catalog No. 501945463
Encompass_Preferred
Manufactured for type 1 class a Borosilicate glass. Ideal for growing, storing, and mixing of aerobic and anaerobic cultures. Graduated to approximate volumes.. MADE IN THE USA Size: 50ml Closure: 18 Dimensions: 50x105mm Stainless steel closure or Kaput closure sold separately
Catalog No. 50-194-5463 Supplier Bellco Glass, Inc. Supplier No. 251100050
Manufactured for type 1 class a Borosilicate glass. Ideal for growing, storing, and mixing of aerobic and anaerobic cultures. Graduated to approximate volumes.. MADE IN THE USA Size: 50ml Closure: 18 Dimensions: 50x105mm Stainless steel closure or Kaput closure sold separately