Learn More
Bellco Glass, Inc. Delong Roux Cult Bottle,1200mL Schott Blank
SDP
Catalog No. 501945741
Encompass_Preferred
Ideal for large area mono-layer cell culture growth. Offset neck prevents culture from entering neck area. Working volume is approximately one half total volume. DeLong style neck is designed for use with 38mm stainless steel closure. ~ DIMENSIONS: 122mm OD X 56mm W x 290mm OAH 2 per case
Catalog No. 50-194-5741 Supplier Bellco Glass, Inc. Supplier No. 563012000
Ideal for large area mono-layer cell culture growth. Offset neck prevents culture from entering neck area. Working volume is approximately one half total volume. DeLong style neck is designed for use with 38mm stainless steel closure. ~ DIMENSIONS: 122mm OD X 56mm W x 290mm OAH 2 per case