Learn More
Strem, An Ascensus Company 10g. Deloxan MP Metal Scavengers (Thio-Functionalized Polysiloxane).
SDP
Catalog No. NC1769959
Encompass
CAS# . 10g. Deloxan MP Metal Scavengers (Thio-Functionalized Polysiloxane). . Molecular Weight: . Molecular Formula: . Color/form: Abrasion-proof spheres; Water content: 50-70% Strem# 14-1871. http://www.strem.com/catalog/v/14-1871/
Catalog No. NC1769959 Supplier Strem, An Ascensus Company Supplier No. 14187110G
CAS# . 10g. Deloxan MP Metal Scavengers (Thio-Functionalized Polysiloxane). . Molecular Weight: . Molecular Formula: . Color/form: Abrasion-proof spheres; Water content: 50-70% Strem# 14-1871. http://www.strem.com/catalog/v/14-1871/