Learn More
Medchemexpress LLC HY-100180 5mg Medchemexpress, Delpazolid CAS:1219707-39-7 Purity:>98%
SDP
Catalog No. 501931737
Encompass_Preferred
Medchemexpress, HY-100180 5mg Delpazolid CAS:1219707-39-7 Delpazolid is a novel oxazolidinone antibiotic agent which can inhibit the growth of MSSA and MRSA with a MIC 90 of 2 μg/mL for both of them. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1737 Supplier Medchemexpress LLC Supplier No. HY1001805MG
Medchemexpress, HY-100180 5mg Delpazolid CAS:1219707-39-7 Delpazolid is a novel oxazolidinone antibiotic agent which can inhibit the growth of MSSA and MRSA with a MIC 90 of 2 μg/mL for both of them. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.