Learn More
Medchemexpress LLC Delphinidin-3-O-galactoside chloride | 28500-00-7 | 1 MG
SDP
Catalog No. 5000418921
Encompass_Preferred
Delphinidin-3-O-galactoside (chloride) is an anthocyanin extracted from wheat flour. It is utilized in research for its antioxidant and antimicrobial properties.
  • Anthocyanin extracted from wheat flour.
  • Used in antioxidant research.
  • Used in antimicrobial research.
Catalog No. 50-004-18921 Supplier Medchemexpress LLC Supplier No. HYN66061MG
Delphinidin-3-O-galactoside (chloride) is an anthocyanin extracted from wheat flour. It is utilized in research for its antioxidant and antimicrobial properties.
  • Anthocyanin extracted from wheat flour.
  • Used in antioxidant research.
  • Used in antimicrobial research.