Learn More
Enzo Life Sciences Delphinidin chloride (high purity) (10mg). CAS: 528-53-0
SDP
Catalog No. 502009055
Encompass_Preferred
Anthocyanidin with antioxidant effect found in pigmented fruits and vegetables. Shown to inhibit angiogenesis and endothelial cell apoptosis by stimulating nitric oxide (NO) production. Inhibits solar radiation (UVB)-mediated oxidative stress, reducing DNA damage. Delphinidin induces necrosis in hepatocellular carcinoma cells with no caspase activation. Purity: ≥97% (HPLC). Solubility: Soluble in water or DMSO. Long Term Storage: -20°C.
Catalog No. 50-200-9055 Supplier Enzo Life Sciences Supplier No. ALX385028M010
Anthocyanidin with antioxidant effect found in pigmented fruits and vegetables. Shown to inhibit angiogenesis and endothelial cell apoptosis by stimulating nitric oxide (NO) production. Inhibits solar radiation (UVB)-mediated oxidative stress, reducing DNA damage. Delphinidin induces necrosis in hepatocellular carcinoma cells with no caspase activation. Purity: ≥97% (HPLC). Solubility: Soluble in water or DMSO. Long Term Storage: -20°C.