Learn More
Medchemexpress LLC Delphinidin chloride | 528-53-0 | 1 MG
SDP
Catalog No. 5000421848
Encompass_Preferred
Delphinidin chloride is an anthocyanin isolated from berries and red wine. It exhibits endothelium-dependent vasodilation and anticancer activity. This compound modulates JAK/STAT3 and MAPK signaling, thereby inducing apoptosis in HCT116 cells. It also acts as a potent inhibitor of EGFR, shutting down downstream signaling cascades. This product is intended for research use only.
  • Exhibits endothelium-dependent vasodilation
  • Demonstrates anticancer activity
  • Modulates JAK/STAT3 and MAPK signaling
  • Induces apoptosis in HCT116 cells
  • Potent inhibitor of EGFR (IC50: 1.3 μM)
  • Purity of 98.0%
Catalog No. 50-004-21848 Supplier Medchemexpress LLC Supplier No. HYN24091MG
Delphinidin chloride is an anthocyanin isolated from berries and red wine. It exhibits endothelium-dependent vasodilation and anticancer activity. This compound modulates JAK/STAT3 and MAPK signaling, thereby inducing apoptosis in HCT116 cells. It also acts as a potent inhibitor of EGFR, shutting down downstream signaling cascades. This product is intended for research use only.
  • Exhibits endothelium-dependent vasodilation
  • Demonstrates anticancer activity
  • Modulates JAK/STAT3 and MAPK signaling
  • Induces apoptosis in HCT116 cells
  • Potent inhibitor of EGFR (IC50: 1.3 μM)
  • Purity of 98.0%