Learn More
Medchemexpress LLC Delphinidin (chloride) | 528-53-0 | 98.0% | 338.70 | C15H11ClO7 | 10 MG
SDP
Catalog No. 5000289442
Encompass_Preferred
Delphinidin (chloride) is an anthocyanidin research standard supplied as a brown-to-black solid. It is provided in a small 10 mg vial with reported purity of 98.0% (NMR) and is intended for analytical and biological research; store sealed at 4°C, away from moisture and light, and prepare fresh solutions as the compound is unstable in solution.
  • Anthocyanidin research standard used for analytical and biological studies.
  • High purity: 98.0% (NMR) for reliable analytical results.
  • Brown-to-black solid appearance, suitable for handling and storage.
  • Recommended storage at 4°C, sealed, away from moisture and light.
  • Unstable in solution; prepare fresh solutions before use.
Catalog No. 50-002-89442 Supplier Medchemexpress LLC Supplier No. HYN240910MG
Delphinidin (chloride) is an anthocyanidin research standard supplied as a brown-to-black solid. It is provided in a small 10 mg vial with reported purity of 98.0% (NMR) and is intended for analytical and biological research; store sealed at 4°C, away from moisture and light, and prepare fresh solutions as the compound is unstable in solution.
  • Anthocyanidin research standard used for analytical and biological studies.
  • High purity: 98.0% (NMR) for reliable analytical results.
  • Brown-to-black solid appearance, suitable for handling and storage.
  • Recommended storage at 4°C, sealed, away from moisture and light.
  • Unstable in solution; prepare fresh solutions before use.